Sleeved Biopsy Cassettes, Dual Hinge Design, Separate Lids, Globe Scientific
Supplier: Globe Scientific
1093SN 1093SH 12093SW 1093SP 1093ST 1093SGR 1093SY
10026-984CS 176.2 USD
10026-984 89800-694 10026-986 10026-982 10026-988 10026-976 10026-978
Sleeved Biopsy Cassettes, Dual Hinge Design, Separate Lids, Globe Scientific
Embedding Cassettes Biopsy Processing Cassettes
Sleeved biopsy cassettes, dual hinge design, separate lids, globe scientific
  • Pre-filled cassette sleeves enable efficient workflow
  • Excellent print quality and readability
  • Ideal for text or barcode printing
  • Resistant to histological solvents and decalcifying solutions
  • Cassettes keep samples safely submerged in liquids
  • Wide slots ensure excellent fluid exchange and proper drainage
  • Feature a 45° angled area for optimal printing
These cassettes assist in keeping samples safely submerged in liquids. 1 mm square openings ensure excellent fluid exchange and proper drainage.
Ordering information: Cassettes are packaged 75/Sleeve, 10 Sleeves/Case. Lids are packaged 75/Bag, 10 Bags/Case.