Would you like to log in again?
Taped Routine Cassettes with Attached Lids, Globe Scientific
Supplier: Globe Scientific
1094TG 1094TW 1094TH 1094TY 1094TGR 1094TB 1094TT 1094TN 1094TP 1094TA 1094TL
One-piece design allows the lids to be opened and closed repeatedly without the lid detaching from the base
Wide slots ensure excellent fluid exschange and proper drainage. The large thumb tab featured on the front of the cassette for easy opening
The lid securely locks into the base and keeps samples safe and secure. The cassettes also feature a 35° angled area for optimal printing. High Density Acetal Polymer material.
Ordering information: Packaged 40/Taped Stack, 25 Stacks/Case.
ORDER
Expand All / Collapse All
Color Supplier No. VWR Catalog Number Unit Price Quantity
Aqua 1094TA 10026-992 Case of 1,000 Retrieving Restricted
Blue 1094TB 10026-994 Case of 1,000 Retrieving Restricted
Green 1094TG 10026-996 Case of 1,000 Retrieving Restricted
Gray 1094TGR 10026-958 Case of 1,000 Retrieving Restricted
Peach 1094TH 10026-960 Case of 1,000 Retrieving Restricted
Lilac 1094TL 10026-962 Case of 1,000 Retrieving Restricted
Orange 1094TN 10026-964 Case of 1,000 Retrieving Restricted
Pink 1094TP 10026-998 Case of 1,000 Retrieving Restricted
Tan 1094TT 10027-000 Case of 1,000 Retrieving Restricted
White 1094TW 10027-002 Case of 1,000 Retrieving Restricted
Yellow 1094TY 10027-004 Case of 1,000 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty