Would you like to log in again?
CD-14, mouse recombinant, BioVision
Supplier: BioVision
4938-10
10007-334
CD-14, mouse recombinant, BioVision
90-95% Pure recombinant active mouse CD-14
Inhibits binding of LPS to CD14. Up to 5µg/ml mouse CD14 inhibit binding of FITC-LPS (0.5µg/ml) to mouse CD14+ CHO transfectants
CD14 acts as a receptor for endotoxin (LPS), is expressed strongly on monocy¬tes, macrophages and weekly at surface of neutrophiles. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor LPS-LBP (lipopolysaccharide binding protein)-complexes. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids, acts at physiological concentrations as LPS agonist and has at higher concentrations an antagonizing effect in cell activation. The myeloid differentiation antigen CD14 acts as the major receptor for bacterial LPS. The dominant form of the recombinant wild type CD14 is the 50 kDa protein. This is a His-tagged protein.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty