>95% Pure active Recombinant human LIF.
The ED₅₀ determined by the M1 cell differentiation assay is < 0.01 ng/ml, corresponding to a specific activity of 100MIU/mg.
Leukemia Inhibitory Factor (LIF) is a lymphoid factor that promotes long-term maintenance of embryonic stem cells by suppressing spontaneous differentiation. LIF has several functions such as cholinergic neuron differentiation, control of stem cell pluripotency, bone & fat metabolism, mitogenesis of factor dependent cell lines & promotion of megakaryocyte production in vivo. Human and mouse LIF exhibit a 78% identity in its amino acid sequence. Human LIF is as active on human cells as is it is on mouse cells, though mouse LIF is about 1000 fold less active on human cells, than human LIF. Recombinant human LIF produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 180 amino acids and having a molecular mass of 19.7 kDa.