Would you like to log in again?
Angiostatin K1-3, human recombinant, BioVision
Supplier: BioVision
4920-1000 4920-100 4920-20
10007-314EA 2039.4 USD InStock
10007-314 10007-312 10006-956
Angiostatin K1-3, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Recombinant human Angiostatin K1-3.
The activity is assayed on anti-proliferation and anti-migration of endothelial cells in vitro and anti-angiogenesis in vivo. The specific activity of anti-migration of endothelial cells in vitro is 0.55×10⁵ Units/mg.
Ang K1-3 is a single, non-glycosylated polypeptide chain containing 259 amino acids. It represents a proteolytic fragment of plasminogen containing the first three kringle structures. A specific inhibitor of endothelial cell growth and angiogenesis. Displays increased inhibitory activity (ED₅₀ =70 nM) relative to kringles 1-4 (ED₅₀ = 135 nM). Ang K1-3 reduces endothelial cell proliferation and acts as a potent inhibitor of angiogenesis and tumor growth.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty