Would you like to log in again?
Gremlin-1, human recombinant, BioVision
Supplier: BioVision
7159-10 7159-50
10009-284EA 87.55 USD InStock
10009-284 10009-286
Gremlin-1, human recombinant, BioVision
Proteins and Peptides
>90% Pure active Human Recombinant Gremlin-1.
Determined by its ability to inhibit BMP-4 induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The ED50 for this effect is 0.07-0.11 μg/ml.
Gremlin-1 (isoform-1) belongs to a group of diffusible proteins which bind to ligands of the TGF-β family and regulate their activity by inhibiting their access to signaling receptors. The interplay between TGF-β ligands and their natural antagonists has major biological significance during development processes, in which cellular response can vary considerably depending upon the local concentration of the signaling molecule. Gremlin is highly expressed in the small intestine, fetal brain, and colon and lower expression in brain, prostate, pancreas and skeletal muscle. Gremlin-1 regulates multiple functions in early development by specifically binding to and inhibiting the function of BMP-2, -4, and -7. It also plays a role in carcinogenesis and kidney branching morphogenesis. Recombinant Gremlin-1 is an 18.3 kDa protein containing 160 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty