Would you like to log in again?
BMP-11/GDF-11, human recombinant, BioVision
Supplier: BioVision
7155-10 7155-50
10008-828 10008-830
BMP-11/GDF-11, human recombinant, BioVision
>98% Pure active Human Recombinant GDF-11.
Determined by its ability to inhibit induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The ED50 for this effect is 0.08-0.10 μg/ml.
GDF-11 is a myostatin-homologous protein that acts as an inhibitor of nerve tissue growth. GDF-11 has been shown to suppress neurogenesis through a myostatin-like pathway, which involves arrest of progenitor cell cycle in the G1 phase. Similarities between myostatin and GDF-11, which are 90% identical in their amino acid sequence, suggests that the regulatory mechanisms responsible for maintaining proper tissue size during neural and muscular development might be the same. Recombinant human GDF-11 is a 25.0 kDa disulfide-linked homodimer containing two 109 amino acid polypeptide chains. It is highly homologous to myostatin/GDF-8 sharing 90% amino acid sequence identity.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty