Would you like to log in again?
BMP-9/GDF-2, human recombinant, BioVision
Supplier: BioVision
7154-10 7154-50
10008-824EA 216.3 USD InStock
10008-824 10008-826
BMP-9/GDF-2, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Human Recombinant GDF-2.
Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 0.5-1.9 ng/ml.
GDF-2 belongs to the TGF-β cytokine family whose members play an important role during prenatal development and postnatal growth, remodeling and maintenance of a variety of tissues and organs. GDF-2 is expressed mainly in non-parenchymal cells of the liver, but is also found in other various cells and tissues. GDF-2 can signal through the ALK1 receptor and has been implicated in a number of physiologic events. These include regulation of the hepatic reticulo-endothelial system, glucose homeostasis, and iron homeostasis, as well as the inhibition of angiogenesis. Recombinant human GDF-2 is a 24.1 kDa disulfide linked homodimeric protein consisting of two 110 amino acid polypeptide chains.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty