Would you like to log in again?
GCP-2 (CXCL6), human recombinant, BioVision
Supplier: BioVision
7153-50 7153-10
10009-246EA 592.25 USD InStock
10009-246 10009-244
GCP-2 (CXCL6), human recombinant, BioVision
Proteins and Peptides
>98% Pure active Human Recombinant GCP-2 (CXCL6).
Determined by its ability to chemoattract human neutrophils using a concentration range of 10.0-50.0 ng/ml.
GCP-2 is a connective tissue derived CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. GCP-2 selectively attracts neutrophils and has also been shown to exert angiogenic activity. Human GCP-2 is cross-react on murine cells. Recombinant human GCP-2 is a 7.9 kDa protein containing 72 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty