Would you like to log in again?
FGF-17, human recombinant, BioVision
Supplier: BioVision
7150-10 7150-50
10009-232EA 149.35 USD InStock
10009-232 10009-234
FGF-17, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Human Recombinant FGF-17.
Determined by a cell proliferation assay using balb/c 3T3 cells. The expected ED50 is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^5 units/mg.
FGF-17 is a heparin binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-17 signals through the FGFR 1c, 2c, 3c, and 4. FGF-17 signals induction and patterning of embryonic brain. Recombinant human FGF-17 is a 22.7 kDa protein consisting of 195 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty