Human Recombinant FGF6 (from E. coli)
Supplier: BioVision
7148-10 7148-50
10009-224EA 169.28 USD
10009-224 10009-226
Human Recombinant FGF6 (from E. coli)
>95% Pure active Human Recombinant FGF-6.
Determined by the dose dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors. The expected ED50 is ≤ 0.5 ng/ml corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
FGF-6 is a secreted heparin binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-6 is expressed in leukemia cell lines with platelet megakaryocytic differentiation potential. It signals through FGFR 1c, 2c, and 4. Recombinant human FGF-6 is an 18.7 kDa protein consisting of 168 amino acid residues.