Would you like to log in again?
FGF-5, human recombinant, BioVision
Supplier: BioVision
7147-50 7147-10
10009-222EA 293.55 USD InStock
10009-222 10009-220
FGF-5, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Human Recombinant FGF-5.
Determined by the dose dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors. The expected ED50 is ≤ 0.5 ng/ml corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
FGF-5 is a secreted heparin binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-5 binds to FGFR 1c and 2c, and plays a regulatory role in the hair growth cycle. Recombinant human FGF-5 is a 27.6 kDa protein consisting of 252 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty