Would you like to log in again?
sFas Receptor, human recombinant, BioVision
Supplier: BioVision
7141-50 7141-10
10008-766EA 767.35 USD InStock
10008-766 10008-764
sFas Receptor, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Human Recombinant sFAS Receptor.
ED50 was determined by its ability to inhibit the cytotoxicity of Jurkat cells using a concentration range of 10-15 μg/ml in the presence of 2ng/ml of hFasL.
Fas and Fas Ligand (FasL) belong to the TNF superfamily and are type I and type II transmembrane proteins, respectively. Binding of FasL to Fas triggers apoptosis in Fas-bearing cells. The mechanism of apoptosis involves recruitment of pro-caspase 8 through an adaptor molecule called FADD followed by processing of the pro-enzyme to active forms. These active caspases then cleave various cellular substrates leading to the eventual cell death. sFasR is capable of inhibiting FasL induced apoptosis by acting as a decoy receptor that serves as a sink for FasL. The full length Fas (receptor) is a 319 amino acid type I transmembrane protein, which contains a 157 amino acid extracellular domain, a 17 amino acid transmembrane domain, and 145 amino acid cytoplasmic domain. Recombinant human soluble Fas (sFas Receptor) is a 157 amino acid polypeptide (17.6 kDa) corresponding to the TNFR homologous cysteine rich extracellular domain Fas.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty