>98% Pure active Murine Recombinant Eotaxin-2.
Determined by its ability to chemoattract murine lymphocytes using a concentration range of 10.0-100.0 ng/ml.
Eotaxin-2 is a CC chemokine that signals through the CCR3 receptor. It is produced by activated monocytes and T lymphocytes. Eotaxin-2 selectively chemoattracts cells expressing CCR3 including eosinophils, basophils, Th2 T cells, mast cells, and certain subsets of dendritic cells. Additionally, Eotaxin-2 inhibits the proliferation of multipotential hematopoietic progenitor cells. The mature protein, which also includes a C-terminal truncation, contains 78 amino acid residues (92 a.a. residues for the murine homolog, without C-terminal truncation). Recombinant murine Eotaxin-2 is a 10.3 kDa protein containing 93 amino acid residues.