Would you like to log in again?
BMPR1A, human recombinant, BioVision
Supplier: BioVision
4881-10 4881-1000
10007-272EA 339.9 USD InStock
10007-272 10007-274
BMPR1A, human recombinant, BioVision
Proteins and Peptides
>90% Pure active Recombinant human BMPR1A.
Measured by its ability to inhibit recombinant human BMP-2 induced alkaline phosphatase production by C2C12 myogenic cells. The ED₅₀ for this effect is typically 1-3 µg/ml in the presence of 500 ng/ml of recombinant human BMP-2.
The bone morphogenetic protein (BMP) receptors are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the Activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kDa and type II receptors of about 70-80 kDa. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. BMPR1A Human Recombinant extracellular domain produced in baculovirus is a monomeric, glycosylated, polypeptide chain fused with 6xHis tag at C-terminus and having a molecular mass of 23 kDa.
The BMR1A is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty