Human Recombinant GMFG (from E. coli)
Supplier: BioVision
4879-1000 4879-10
10007-264EA 315.22 USD
10007-264 10007-266
Human Recombinant GMFG (from E. coli)
>90% Pure Recombinant human GMF-gamma.
GMFG is a hematopoietic-specific protein that mediates the pluripotentiality and lineage commitment of human hematopoietic stem cells. Glia maturation factor gamma is a cytokine-responsive protein in EPO-induced and G-CSF-induced hematopoietic lineage development. Glia maturation factor also acts as a Nerve Growth Factor in nervous system development, angiogenesis and immune function. GMFG possesses hematopoietic tissue-specific gene expression, a promoter concentrated with high-score hematopoiesis-specific transcription factors, and molecular coevolution with a rudimentary blood/immune system. Glia Maturation Factor-Gamma (GMF-Gamma) Human Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 142 amino acids and having a total molecular mass of 16.8 kDa. Glia Maturation Factor-Gamma, GMF-Gamma, Human Recombinant is purified by proprietary chromatographic techniques.