Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human CellExp™ Cystatin C, Human Recombinant, BioVision

Supplier: BioVision
4878-10 4878-50
>95% Pure Recombinant human Cystatin C.

The inhibitory function of Cystatin C on papin’s protease activity was measured by a colorimetric assay using L-BAPA as substrate. IC50 value was measured at 5 to 20 µg/mL (0.3 to 1.5 µM) with a range of 1.56 µg/mL to 50 µg/mL Cystatin C in presence of 0.55 µM papain and 0.44 µM L-BAPA.

Xeno-free Cystatin C HuXp (Cys-C) is expressed in human 293 cells as a monomer with an apparent molecular mass of 12 to 13 kDa. Native Cys-C in human urine is found in two different forms: one with pI 9.2 and the other with pI 7.8 by elimination of small basic peptides or amino acids from the N-terminal end of protein. Cystatin C has been studied for its role in predicting new onset or deteriorating cardiovascular disease. This cytokine is produced in a serum-free, chemically defined media.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 μg 4878-10 10007-260 Each (10µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

50 μg 4878-50 10007-262 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Cystatin C
Protein Synonyms Cystatin-3, Post-gamma-globulin, Gamma-trace neuroendocrine basic polypeptide
Protein/Peptide Type Recombinant
Species Human
Source 293 cells
Purity ≥95%
Endotoxin Content <0.1 ng/μg
Molecular Weight 12-13 kDa
Formulation Lyophilized in PBS
Reconstitution Instructions Reconstitute in sterile PBS.
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty