Human Recombinant Scd22 (from CHO Cells)
Supplier: BioVision
7123-10 7123-50
10009-154EA 963.05 USD
10009-154 10009-152
Human Recombinant Scd22 (from CHO Cells)
>98% Pure active Human Recombinant sCD22.
Determined by its ability to inhibit the proliferation of Raji cells. The expected ED50 for this effect is 10-17 μg/ml.
CD22 is a B-lineage restricted 135 kDa glycoprotein whose cell surface expression is limited to resting and activated B lymphocytes. The physiological role of CD22 is still unknown. Targeted disruption of CD22 in mice results in a reduced level of surface IgM on peripheral B cells suggesting a role for CD22 in limiting antigen receptor signaling. CD22 is a member of the Ig gene superfamily that uniquely binds a sialic acid-dependent ligand. Recombinant human CD22 is a soluble 75.0 kDa protein which corresponds to the extracellular domain of CD22.