Mouse Recombinant Cardiotrophin 1 (from E. coli)
Supplier: BioVision
7121-10 7121-50
10009-144EA 245.17 USD
10009-144 10009-146
Mouse Recombinant Cardiotrophin 1 (from E. coli)
>98% Pure active Murine Recombinant Cardiotrophin 1
Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
CT-1 is a member of the IL-6 family of cytokines which also includes LIF, CNTF, OSM (Oncostatin M), IL-11, IL-6 and possibly NT-1/ BSF-3. CT-1 is a pleiotropic cytokine which is expressed in various tissues including the adult heart, skeletal muscle, ovary, colon, prostate and fetal lung and signals through the LIF receptor and the gp130 receptor subunit. CT-1 has the ability to induce cardiac myocyte hypertrophy, and enhances the survival of cardiomyocyte and different neuronal populations. Biologically active human CT-1 is synthesized as a 201 amino acid polypeptide lacking a hydrophobic N-terminal secretion signal sequence. Recombinant murine Cardiotrophin-1 is a 21.3 kDa protein consisting of 202 amino acid residues.