Would you like to log in again?
IL-36γ, Human Recombinant, BioVision
Supplier: BioVision
7111-50 7111-10
10009-106EA 1024.85 USD InStock
10009-106 10009-104
IL-36γ, Human Recombinant, BioVision
Proteins and Peptides
>98% Pure active Human Recombinant IL-36γ.
Measured by its ability to induce IL-8 production by human PBMCs
The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-36 α (IL-1F6), β (IL-1F8) and γ (IL-1F9). IL-36γ is highly expressed in psoriatic plaques and in tissues containing epithelial cells. IL-36γ signals through the IL-1Rrp2 (IL-1R6) receptor, which is primarily expressed on certain dendritic cells. The interaction of the IL-1Rrp2 receptor with IL-36 ligands induces dendritic cell maturation and activation. IL-36γ also functions as an agonist of NF-kappaB, and can stimulate the inflammatory response in bronchial epithelial cells. Recombinant human IL-36γ is a 17.0 kDa protein containing 152 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty