Human Recombinant Il 17B (from E. coli)
Supplier: BioVision
7106-50 7106-10
10009-084EA 157.61 USD
10009-084 10009-086
Human Recombinant Il 17B (from E. coli)
>98% Pure active Human Recombinant IL-17B.
Determined by its ability to induce IL-8 production in human PBMCs using a
concentration range of 10.0-100.0 ng/ml. Please note: Results may vary with PBMC donors.
IL-17B is a disulfide-linked homodimer of two 161 amino acid polypeptide chains. It belongs to the IL-17 family of structurally-related cytokines that share a highly conserved C-terminal region but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17B is expressed by T-cells and has been shown to stimulate release of TNF-α and IL-1β from cells of the monocyte lineage. Recombinant human IL-17B is a 36.6 kDa nondisulfide-linked homodimer of two 161 amino acid polypeptide chains.