Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human CellExp™ IL-4 Rα, Human Recombinant, BioVision

Supplier: BioVision
7101-50 7101-10
>98% Pure active Human Recombinant Human cell expressed IL-4 Rα.

Determined by its ability to inhibit the IL-4-dependent proliferation of human TF-1 cells. The expected ED50 is ≤ 5.0 ng/ml (in the presence of 0.5 ng/ml of hIL-4), corresponding to a specific activity of ≥ 2 x 105 units/mg.

IL-4 can signal through type I and type II receptor complexes, which share a common gamma chain (γc). The type I receptor contains in addition to the γc an IL-4Rα subunit, whereas the type II receptor contains the IL-13Rα. The secreted extracellular domain of IL-4Rα, called sIL-4Rα, binds IL-4 and antagonizes its activity. It plays an important role in regulating the differentiation of naïve CD4+ T cells and class switching to IgG1 and IgE. Recombinant human sIL-4Rα is a 209 amino acid protein which corresponds to the entire extracellular domain of IL-
4Rα.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 μg 7101-10 10009-064 Each (10µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

50 μg 7101-50 10009-066 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name IL4 Receptor
Protein Synonyms soluble IL-4 receptor alpha
Protein/Peptide Type Recombinant
Species Human
Source HEK293 cells
Purity ≥98%
Endotoxin Content < 0.1 ng/μg of protein (<1EU/μg).
Molecular Weight ~24 kDa
Formulation Sterile filtered through a 0.2 micron filter. Lyophilized from 1x PBS pH 7.0.
Reconstitution Instructions Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex.
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty