Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human CellExp™ TGF-beta 2, Human Recombinant, BioVision

Supplier: BioVision
6480-50 6480-10
>95% Pure HEK 293 cells expressed Human recombinant TGF-beta 2

ED₅₀ is typically 0.1 to 0.5 ng/mL. The specific activity was determined by the dose-dependent inhibition of IL-4 induced proliferation of mouse HT-2 cells (BALB/c spleen activated by sheep erythrocytes in the presence of IL-2).

Transforming growth factor betas (TGFβ) mediate many cell-cell interactions that occur during embryonic development. Three TGFβs have been identified in mammals. TGF Beta 1, TGF Beta 2 and TGF Beta 3 are each synthesized as precursor proteins that are very similar in that each is cleaved to yield a 112 amino acid polypeptide that remains associated with the latent portion of the molecule. TGF-beta 1 is involved in hematopoiesis and endothelial differentiation.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 µg 6480-10 10008-728 Each (10µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

50 µg 6480-50 10008-730 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name TGF beta TGF beta 2
Protein Synonyms transforming growth factor beta-2, TGF-beta2, TGFB-2, G-TSF, TGF-beta-2, BSC-1 cell growth inhibitor TGF-beta2, transforming growth factor beta-2, TGFB-2, G-TSF, TGF-beta-2, BSC-1 cell growth inhibitor
Protein/Peptide Type Recombinant
Species Human
Source 293 cells
Gene ID 7042
Purity > 95%
Endotoxin Content < 1.0 EU per 1 μg of protein
Molecular Weight 25 kDa
Formulation Lyophilized from 50 mM sodium acetate, pH 4.5 and 350 mM NaCl.
Reconstitution Instructions Reconstitute in sterile 4 mM HCl containing 0.1% endotoxin-free recombinant human serum albumin.
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty