Human Recombinant BLMH (from E. coli)
Supplier: BioVision
AB168022-100UG
10008-506EA 781.41 USD
10008-506
Human Recombinant BLMH (from E. coli)
Proteins and Peptides
>90% Pure Active Recombinant Human BLMH
Specific activity: > 1,000 pmole/min/µg. Measured by the hydrolysis of Met-AMC at pH 7.5, at 37°C.
BLMH is affiliate to the papain superfamily of the cysteine protease and the peptidase C1 family. BLMH is a cytoplasmic cysteine peptidase usually found as a homohexamer. The standard physiological role of BLMH has not been determined, but it shields normal and malignant cells from the glycopeptide antitumor drug BLM. BLMH catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxyamide bond of its B-aminoalaninamide moiety and in addition demonstrates general aminopeptidase activity.