Human Recombinant Sord (from E. coli)
Supplier: BioVision
AB167975-100UG
10008-498EA 781.44 USD
10008-498
Human Recombinant Sord (from E. coli)
Proteins and Peptides
>90% Pure Active Recombinant Human SORD
Specific activity is > 0.2 units/mg
SORD, also known as L-iditol 2-dehydrogenase or SORD1, is a 357 amino acid member of the zinc containing alcohol dehydrogenase family. It is widely expressed with highest expression in kidney and in the lens of the eye. SORD enzymatically catalyzes the zinc-dependent interconversion of polyols, such as sorbitol and xylitol, to their respective ketoses.