Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human CellExp™ IL-17F, Human Recombinant, BioVision

Supplier: BioVision
6469-50 6469-10
>95% Pure HEK 293 cells expressed Human recombinant IL-17F

ED₅₀ is typically 100 to 500 ng/mL. The specific activity was determined by the dose-dependent induction of IL-6 secretion from NHDF Adult fibroblasts.

Interleukin-17 is a proinflammatory cytokine produced by activated T cells. IL-17 regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL17 can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-17 are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis. IL-17F is a member of the IL-17 family of structurally-related cytokines. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17F stimulates proliferation and activation of T-cells and PBMCs. IL-17F also regulates cartilage matrix turnover and inhibits angiogenesis.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 µg 6469-10 10008-692 Each (10µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

50 µg 6469-50 10008-694 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name IL17F
Protein Synonyms Interleukin-17 AF, Interleukin-17 A/F, IL17 A/F, IL-17 A/F, IL17A/F, IL17AF, IL-17A/F, IL-17 AF IL-17A/F, IL-17 AF, IL17A/F, IL17 A/F, IL17AF, IL-17 A/F, Interleukin-17 AF, Interleukin-17 A/F
Protein/Peptide Type Recombinant
Species Human
Source 293 cells
Purity > 95%
Endotoxin Content < 1.0 EU per 1 μg of protein
Molecular Weight 38 kDa
Formulation Lyophilized in PBS.
Reconstitution Instructions Reconstitute in sterile PBS containing 0.1% endotoxin-free recombinant human serum albumin.
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping