Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human CellExp™ IL-17A, Human Recombinant, BioVision

Supplier: BioVision
6468-10 6468-50
>95% Pure HEK 293 cells expressed Human recombinant IL-17A

ED₅₀ is typically 5 to 25 ng/mL. The specific activity was determined by the dose-dependent induction of IL-6 secretion from NDHF Adult fibroblasts.

Interleukin-17 is a proinflammatory cytokine produced by activated T cells. IL-17 regulates the activities of NF-kappa B and mitogen-activated protein kinases. IL17 can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-17 are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis. The original described IL-17 protein, now known as IL-17A, was identified from a CD4+ T cells DNA library. IL-17 can be induced from primary peripheral blood CD4+ T cells upon stimulation. Supernatant from cells transfected with IL-17 induced IL-16 and IL-18 production and enhanced the surface expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 µg 6468-10 10008-688 Each (10µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

50 µg 6468-50 10008-690 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name IL17A
Protein Synonyms CTLA-8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17, IL-17A Cytotoxic T-lymphocyte-associated antigen 8, CTLA-8, IL-17, IL-17A
Protein/Peptide Type Recombinant
Species Human
Source 293 cells
Purity > 95%
Endotoxin Content < 1.0 EU per 1 μg of protein
Molecular Weight 30 to 35 kDa
Formulation Lyophilized in 10 mM Tris, pH 7.4 and 250 mM NaCl.
Reconstitution Instructions Reconstitute in sterile PBS containing 0.1% endotoxin-free recombinant human serum albumin.
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping