Human Recombinant PIN4 (from E. coli)
Supplier: BioVision
6387-100
10008-496EA 472.69 USD
10008-496
Human Recombinant PIN4 (from E. coli)
>85% Pure Active Recombinant Human PIN4
Specific activity is > 300 nmoles/min/mg
Peptidyl-prolyl cis-trans isomerase NIMA-interacting 4 (PIN4) is a peptidyl-prolyl cis/trans isomerase (PPIase) which interacts with NIMA and is vital for cell cycle regulation. PIN4 has 2 different isoforms: PAR14 and PAR17. Furthermore, Pin4 protein binds to double-stranded DNA under physiological salt conditions.