Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant DHFR, BioVision

Supplier: BioVision
6382-100
>95% Pure Active Recombinant Human DHFR

Specific activity is 1.5 - 2.5 units/ml and was obtained by measuring the oxidation of NADPH in absorbance at 340 nm during reaction.

DHFR, also known as Dihydrofolate reductase, is an enzyme that reduces dihydrofolic acid to tetrahydrofolic acid, using NADPH as electron donor, which can be converted to tetrahydrofolate cofactors used in 1-carbon transfer chemistry. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia. Human dihydrofolate reductase has been used in a study to investigate the stable expression of green fluorescent protein and the targeted disruption of thioredoxin peroxidase-1 gene in Babesia bovis. Human dihydrofolate reductase has also been used in a study to investigate the structural analysis of human dihydrofolate reductase as a binary complex.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg 6382-100 10008-486 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Dhfr
Protein/Peptide Type Recombinant
Species Human
Source E. coli
Purity ≥95%
Endotoxin Content < 1.0 EU per 1 μg of protein (determined by LAL method)
Molecular Weight 23.6 kDa
Formulation 1 mg/ml solution in 20 mM Tris-HCl buffer (pH 8.0) containing 0.1 M NaCl, 2 mM DTT and 30% glycerol.
Biological Activity Specific activity is 1.5 - 2.5 units/ml
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty