Recombinant Gor (from E. coli)

Supplier: BioVision
AB167992-100UG
10008-482EA 865.68 USD
10008-482
Recombinant Gor (from E. coli)
Proteins and Peptides
>90% Pure Active Recombinant Bacterial GOR

Specific activity: > 52 unit/ml.

Gor, also known as Glutathione oxido-reductase, belongs to the class-I pyridine nucleotide disulfide oxidoreductase family. The main function of this protein is to maintain high levels of reduced glutathione in the cytosol. With the concomitant oxidation of NADPH, Glutathione reductase transforms oxidized glutathione to the reduced form.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR