Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human CellExp™ IFN-alpha 2a, Human Recombinant, BioVision

Supplier: BioVision
CellExp™
6458-50 6458-10
>95% Pure HEK 293 cells expressed Human recombinant IFN-alpha 2a

ED₅is typically 0.1 to 0.4 ng/mL. The specific activity was determined by the dose-dependent cytotoxicity of the human TF -1 cell line(human erythroleukemic indicator cell line).

Interferons (IFNs) are proteins made and released by host cells in response to the presence of pathogens. They belong to the large class of glycoproteins known as cytokines. IFN-alpha is produced by macrophages and has antiviral activities. IFNs also have other functions: they activate immune cells, such as natural killer cells and macrophages; they increase recognition of infection or tumor cells by up-regulating antigen presentation to T lymphocytes; and they increase the ability of uninfected host cells to resist new infection by virus.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 μg 6458-10 10008-648 Each (10µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

50 μg 6458-50 10008-650 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name IFN IFN alpha 2A
Protein Synonyms IFNA2, Type I interferon, B cell interferon, Leukocyte interferon, IFN-A 2a IFNA2, IFN-A 2a, Type I interferon, Leukocyte interferon, B cell interferon
Protein/Peptide Type Recombinant
Species Human
Source 293 cells
Gene ID 3440
Purity ≥95%
Endotoxin Content < 1.0 EU per 1 μg of protein
Molecular Weight 16 kDa
Formulation Lyophilized from a TBS solution.
Reconstitution Instructions Reconstitute in sterile PBS containing 0.1% endotoxin-free recombinant human serum albumin.
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty