Would you like to log in again?
Human Recombinant FKBP1B, BioVision
Supplier: BioVision
6341-100
10008-374EA 386.25 USD InStock
10008-374
Human Recombinant FKBP1B, BioVision
Proteins and Peptides
>90% Pure Recombinant Active Human FKBP1b.
Specific activity is > 300 nmoles/min/mg
Peptidyl-prolyl cis-trans isomerase FKBP1B (FKBP1B) belongs to the immunophilin protein family which has a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP1B is a cis-trans prolyl isomerase which binds the immunosuppressants FK506 (tacrolimus) and rapamycin (sirolimus). FKBP1B is extremely similar to the FK506-binding protein 1A. The physiological role of FKBP1B is thought to be in excitation-contraction coupling in cardiac muscle.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty