Human Recombinant TNF beta (from E. coli)
6376-50
10008-442EA 414.47 USD
10008-442
Human Recombinant TNF beta (from E. coli)
>90% Pure Active Recombinant Human TNF-beta/LTA
Measured in a cytotoxicity assay using L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D. The ED50 for this effect is <0.3 ng/ml.
TNF-beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. It is highly inducible, secreted, and exists as homotrimeric molecule. It forms heterotrimers with lymphotoxin-beta which anchors lymphotoxin-alpha to the cell surface. It mediates a large variety of inflammatory, immunostimulatory, and antiviral responses. It is also involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis.