CellExp™ Human Recombinant G-CSF (from 293 Cells)
Supplier: BioVision
CellExp™
AB285737-50UG AB285737-10UG
10008-630EA 3186.93 USD
10008-630 10008-628
CellExp™ Human Recombinant G-CSF (from 293 Cells)
Proteins and Peptides
>95% Pure HEK 293 cells expressed Human recombinant G-CSF
ED is typically 0.02 to 0.08 ng/mL. The specific activity was determined by the dose - Dependent stimulation of the proliferation of murine M-NFS-60 cells (Mouse Myeloid Leukemia indicator cell line).
GCSF is a cytokine that controls the production, differentiation, and function of granulocytes. The active protein is found extracellularly. Three transcript variants encoding three different isoforms have been found for the GCSF gene. Granulocyte/macrophage colony-stimulating factors are cytokines that act in hematopoiesis by controlling the production, differentiation, and function of 2 related white cell populations of the blood, the granulocytes and the monocytes-macrophages.