Human Recombinant inantpkm2 (from E. coli)
Supplier: BioVision
6372-100
10008-434EA 356.08 USD
10008-434
Human Recombinant inantpkm2 (from E. coli)
>95% Pure Active Recombinant Human PKM2
Specific activity: >5.0 unit/mg.
Pyruvate kinase is a key enzyme in the glycolytic pathway. The M2 isoenzyme of pyruvate kinase is specifically expressed at high levels in tumor cells, and can be measured in plasma of patients with advanced breast cancer. The marker is useful for measuring disease activity, sensitivity to chemotherapy and recurrence. PKM2 interacting with Opa proteins, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, is required for bacterial pathogenesis.