Would you like to log in again?
Human Recombinant PPP1CA, BioVision
Supplier: BioVision
6370-10
10008-430EA 334.75 USD InStock
10008-430
Human Recombinant PPP1CA, BioVision
Proteins and Peptides
>80% Pure Active Recombinant Human PPP1CA
Specific activity: >3,000 units/mg.
Serine/Threonine-protein phosphatase PP1-alpha catalytic subunit (PPP1CA) is crucial for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. The PPP1CA protein may have a significant role in dephosphorylating substrates such as the postsynaptic density-associated Ca2+/calmodulin dependent protein kinase II.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty