Would you like to log in again?
Human Recombinant PPM1G, BioVision
Supplier: BioVision
6369-50
10008-428EA 365.65 USD InStock
10008-428
Human Recombinant PPM1G, BioVision
Proteins and Peptides
>85% Pure Active Recombinant Human PPM1G
Specific activity: >5,000 units/mg.
PPM1G is part of the PP2C family of Ser/Thr protein phosphatases which are known to be negative regulators of cell stress response pathways. PPM1G is accountable for the dephosphorylation of Pre-mRNA splicing factors, an important factor for the formation of functional spliceosome. PPM1G regulates cell cycle progression. PPM1G mediates histone dephosphorylation/exchange in response to DNA damage or checkpoint recovery in higher eukaryotes. The degradation of p21/WAF1 induced by PPM1G is mediated in a proteasome-dependent manner. Protein phosphatase 1G regulates assembly and function of the beta-catenin degradation complex.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty