Human Recombinant G6Pd (from E. coli)
Supplier: BioVision
AB167987-100UG
10008-424EA 758.43 USD
10008-424
Human Recombinant G6Pd (from E. coli)
Proteins and Peptides
>90% Pure Active Recombinant E.Coli G6PD
Specific activity is 8-10 units/ml obtained by measuring the increase of NADPH in absorbance at 340 nm resulting from the reduction of NAD or NADP.
Glucose-6-phosphate dehydrogenase is the rate-limiting enzyme of the pentose phosphate pathway, a metabolic pathway that supplies reducing energy to cells by maintaining the level of NADPH. G6PD converts glucose-6-phosphate into 6-phosphoglucono-delta-lactone and simultaneously produce NADPH. The NADPH in turn maintains the level of glutathione in these cells that helps protect the red blood cells against oxidative damage. G6PD deficiency cause acute hemolytic anemia.