Would you like to log in again?
Human Recombinant AKR7A2, BioVision
Supplier: BioVision
6335-100
10008-362EA 419.21 USD InStock
10008-362
Human Recombinant AKR7A2, BioVision
Proteins and Peptides
>90% Pure Recombinant Active Human AKR7A2
Specific activity is approximately 0.25 – 0.3 units/mg.
Aldo-keto reductases, such as AKR7A2, are involved in the detoxification of aldehydes and ketones. This protein can reduce the dialdehyde protein-binding form of aflatoxin B1 (AFB1) to the nonbinding AFB1 dialcohol. It may be involved in protection of liver against the toxic and carcinogenic effects of AFB1, a potent hepatocarcinogen. Also, it has been proposed previously to catalyze the NADPH-dependent reduction of succinic semialdehyde (SSA) to Gamma-Hydroxybutyrate in human brain.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty