Would you like to log in again?
Human Recombinant AKR7A3, BioVision
Supplier: BioVision
6334-50
10008-360EA 417.15 USD InStock
10008-360
Human Recombinant AKR7A3, BioVision
Proteins and Peptides
>95% Pure Recombinant Active Human AKR7A3
Specific activity is approximately > 0.1 units/mg
AKR7A3, is a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. It takes part in the detoxification of aldehydes and ketones. AKR7A3 reduces the dialdehyde protein-binding form of aflatoxin B1 (AFB1) to the non-binding AFB1 dialcohol. AKR7A3 participates in protection of liver against the toxic and carcinogenic effects of AFB1, a potent hepatocarcinogen.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty