Would you like to log in again?
Human Recombinant Beta-Enolase, BioVision
Supplier: BioVision
6364-50
10008-420EA 417.15 USD InStock
10008-420
Human Recombinant Beta-Enolase, BioVision
Enzymes
>95% Pure Active Recombinant Human Beta-Enolase
Specific activity: >1.5 units/ml.
Beta-enolase, also known as ENO3, is one of the three enolase isoenzymes found in mammals. This isoenzyme, a homodimer, is found in skeletal muscle cells in the adult. ENO3 play a role in converting phosphoglyceric acid to phosphenolpyruvic acid in the glycolytic pathway. Mutations in its gene can be associated with metabolic myopathies that may result from decreased stability of the enzyme. Recombinant human ENO3 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty