Would you like to log in again?
E. Coli Recombinant TRXB, BioVision
Supplier: BioVision
6331-100
10008-014EA 355.35 USD InStock
10008-014
E. Coli Recombinant TRXB, BioVision
Proteins and Peptides
>90% Pure Recombinant Active E. Coli TRXB
Specific activity is 4-5 units/ml
Thioredoxin reductases are the only known enzymes to reduce thioredoxin. Two classes of thioredoxin reductase have been identified: one class in bacteria and some eukaryotes and one in animals. Both classes are flavoproteins which function as homodimers. TRXB (Thioredoxin reductase) is a ubiquitous enzyme which is involved in many cellular processes such as cell growth, p53 activity, and protection against oxidation stress
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty