Would you like to log in again?
E. Coli Recombinant Thioredoxin 1, BioVision
Supplier: BioVision
6329-50
10008-010EA 334.75 USD InStock
10008-010
E. Coli Recombinant Thioredoxin 1, BioVision
Proteins and Peptides
>85% Pure Recombinant Active E. Coli TRX1
Specific activity is 16-18 A650/min/mg
Thioredoxin (Trx) is a low molecular weight redox protein. Trx contains a redox active disulfide/dithiol group within the conserved Cys-Gly-Pro-Cys active site. It is involved in the first unique step in DNA synthesis. Trx also provides control over a number of transcription factors affecting cell proliferation and death through a mechanism referred to as redox regulation. It has also recently been found to play a role in cell-to-cell communication.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty