Would you like to log in again?
Human Recombinant PPIL3, BioVision
Supplier: BioVision
6326-100
10008-006
Human Recombinant PPIL3, BioVision
>90% Pure Recombinant Active Human PPIL3
Specific activity is > 280 nmoles/min/mg
Peptidyl-prolyl cis-trans isomerase-like 3 in humans is encoded by the PPIL3 gene. This gene encodes a member of the cyclophilin family. Cyclophilins catalyze the cis-trans isomerization of peptidylprolyl imide bonds in oligopeptides. They have been proposed to act either as catalysts or as molecular chaperones in protein-folding events.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty