Human Recombinant PPIL2 (from E. coli)
6325-50
10008-004EA 472.83 USD
10008-004
Human Recombinant PPIL2 (from E. coli)
>95% Pure Recombinant Active Human PPIL2
Specific activity is > 290 nmoles/min/mg
Peptidyl-prolyl cis-trans isomerase-like 2 in humans is encoded by the PPIL2 gene. This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. This protein interacts with the proteinase inhibitor eglin c and is localized in the nucleus.