Would you like to log in again?
Superoxide Dismutase A (SOD-A), bacterial recombinant, BioVision
Supplier: BioVision
6361-100
10008-414EA 386.25 USD InStock
10008-414
Superoxide Dismutase A (SOD-A), bacterial recombinant, BioVision
Proteins and Peptides
>95% Pure Active Recombinant Bacterial SOD-A.
Specific activity is >350 units/mg
Superoxide dismutase, Mn, also known as sodA, is a member of the iron/manganese superoxide dismutase family. SodA destroys radicals which are normally produced within the cells and which are toxic to biological systems. It works by catalyzing the dismutation of the superoxide radical O2- to O2 and H2O2, which are then metabolized to H2O and O2 by catalase and glutathione peroxidase.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty