Would you like to log in again?
Human Recombinant PRDX 4, BioVision
Supplier: BioVision
6324-100
10008-358EA 355.35 USD InStock
10008-358
Human Recombinant PRDX 4, BioVision
Proteins and Peptides
>90% Pure Recombinant Active Human PRDX 4
Specific activity: approximately 230-310 pmole/min/µg. Enzymatic activity was confirmed by measuring the remaining peroxide after incubation of PRDX 4 and peroxide for 20 min at room temperature.
Peroxiredoxin-4 in humans is encoded by the PRDX4 gene. The protein encoded by this gene is an antioxidant enzyme and belongs to the peroxiredoxin family. It is localized to the cytoplasm. Peroxidases of the peroxiredoxin family reduce hydrogen peroxide and alkyl hydroperoxides to water and alcohol with the use of reducing equivalents derived from thiol-containing donor molecules. This protein has been found to play a regulatory role in the activation of the transcription factor NF-kappa B.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty