Would you like to log in again?
Human Recombinant PRDX 1, BioVision
Supplier: BioVision
6323-100
10008-356EA 314.15 USD InStock
10008-356
Human Recombinant PRDX 1, BioVision
Proteins and Peptides
>90% Pure Recombinant Active Human PRDX 1
Specific activity: approximately 600-670 pmole/min/µg.
Peroxiredoxin-1 in humans is encoded by the PRDX1 gene. This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8 (+) T-cells. It protein may also have a proliferative effect and play a role in cancer development or progression.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty