Would you like to log in again?
Human Recombinant PRDX 6, BioVision
Supplier: BioVision
6322-100
10008-354
Human Recombinant PRDX 6, BioVision
>95% Pure Recombinant Active Human PRDX 6
Specific activity: approximately 95-120 pmole/min/µg.
Peroxiredoxin 6, also known as PRDX 6, is a member of the thiol-specific antioxidant protein family. This protein is a bifunctional enzyme with two distinct active sites. It is involved in redox regulation of the cell and can reduce H2O2 and short chain organic, fatty acid, and phospholipid hydroperoxides. It may play a role in the regulation of phospholipid turnover as well as in protection against oxidative injury.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty