Would you like to log in again?
Human Recombinant PRDX 5, BioVision
Supplier: BioVision
6321-100
10008-352EA 355.35 USD InStock
10008-352
Human Recombinant PRDX 5, BioVision
Proteins and Peptides
>95% Pure Recombinant Active Human PRDX 5
Specific activity: approximately 117-136 pmole/min/µg.
Peroxiredoxin-5, mitochondrial in humans is encoded by the PRDX5 gene. This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in different tissues under normal conditions and during inflammatory processes. It interacts with peroxisome receptor 1. In human cells, it has been shown that PRDX5 can be addressed to mitochondria, peroxisomes, the cytosol, and the nucleus.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty